Ciprofloxacin Hydrochloride, MP Biomedicals™
Ciprofloxacin Hydrochloride, MP Biomedicals™
Ciprofloxacin Hydrochloride, MP Biomedicals™
Ciprofloxacin Hydrochloride, MP Biomedicals™
MP Biomedicals
Ciprofloxacin Hydrochloride, MP Biomedicals™
Very effective against a broad spectrum of bacteria and mycoplasma. Ciprofloxacin Hydrochloride, MP Biomedicals is a fluorinated quinolone antibacterial agent. Typically used for 12–20 days in cell culture media, with the media being changed every 3
Have Questions?
Catalog NumberQuantity
MPB02199020055 g
Catalog number MPB0219902005
Price (SGD)
103.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
103.00
EA
Add to cart
Specifications
Physical FormPowder
CAS86393-32-0
pH3.7 (2% aq. soln.) (literature)
Melting Point313°C to 322°C
Recommended StorageStore at 4°C.
View more
During the proliferation phase of a bacterium, a segmental twisting and untwisting of the chromosomes takes place. An enzyme called DNA gyrase plays a decisive part in this process. Ciprofloxacin inhibits this DNA gyrase in a way that arrests the bacterial metabolism, since vital information can no longer be read from the bacterial chromosome. Ultimately, it affects DNA replication and cell division.
Ciprofloxacin is most commonly used to treat infections of the skin, bone, sinus, lung, abdomen, and bladder. It can also be used to treat some sexually transmitted infections. Additionally, ciprofloxacin can be used after exposure to inhalation of anthrax to reduce the chance of developing an infection.