Check Order Status
Sodium dodecyl sulfate, ≥99.5% alkyl sulfate, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sodium dodecyl sulfate, ≥99.5% alkyl sulfate, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219895780
100 mL
Catalog number
MPB0219895780
Price (SGD)
167.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (SGD)
167.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium dodecyl sulfate, alkyl sulfate
Color
Colorless
CAS
151-21-3
Physical Form
Liquid
Recommended Storage
Store at room temperature.
View more