Check Order Status
Doxycycline hyclate, USP grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Doxycycline hyclate, USP grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219895501
1 g
Catalog number
MPB0219895501
Price (SGD)
149.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
149.00
EA
Add to cart
Specifications
Chemical Name or Material
Doxycycline hyclate
Type
Doxycycline Hyclate
Physical Form
Powder
CAS
24390-14-5
Color
Yellow
View more