Check Order Status
Glutaraldehyde, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Glutaraldehyde, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219859510
10 mL
Catalog number
MPB0219859510
Price (SGD)
142.00
EA
Add to cart
Quantity:
10 mL
Request bulk or custom format
Price (SGD)
142.00
EA
Add to cart
Specifications
Chemical Name or Material
Glutaraldehyde
CAS
111-30-8
Color
Colorless
Density
1.061g/cm
3
at 20°C
Molecular Formula
C5H8O2
View more