Check Order Status
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Have Questions?
Change view
Quantity:
25 mL
100 mL
0.5 L
1 L
Catalog number
MPB0219605525
Price (SGD)
117.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (SGD)
117.00
EA
Add to cart
Specifications
Chemical Name or Material
Dimethyl sulfoxide
CAS
67-68-5
Quantity
25 mL
Physical Form
Liquid
Assay Percent Range
≥99%
View more