Vancomycin Hydrochloride MP Biomedicals
Vancomycin Hydrochloride MP Biomedicals
Vancomycin Hydrochloride MP Biomedicals
Vancomycin Hydrochloride MP Biomedicals
MP Biomedicals
Vancomycin Hydrochloride MP Biomedicals
Have Questions?
Catalog NumberQuantity
MPB02195540011 g
Catalog number MPB0219554001
Price (SGD)
311.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (SGD)
311.00
EA
Add to cart
Specifications
CAS123409-00-7
Quantity1 g
Molecular FormulaC66H257Cl2N9O24
InChI KeyARNOCQCQWLXLRG-UHFFFAOYSA-N
SMILESC.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl
View more