▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Sodium Phosphate Monobasic, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Sodium Phosphate Monobasic, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219550090
500 g
Catalog number
MPB0219550090
Price (SGD)
195.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
195.00
Each
Add to cart
Specifications
Chemical Name or Material
Sodium Phosphate Monobasic
Name Note
Anhydrous
CAS
7558-80-7
Recommended Storage
Room Temperature
Quantity
500 g
View more