Check Order Status
Rifampin, 100.3%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Rifampin, 100.3%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219549005
5 g
Catalog number
MPB0219549005
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
Rifampin
Type
Rifampicin
Physical Form
Powder
CAS
13292-46-1
Color
Orange/Red
View more