Check Order Status
Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Potassium Phosphate Monobasic, 99+%, (Crystalline), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219545301
1 kg
Catalog number
MPB0219545301
Price (SGD)
333.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
333.00
EA
Add to cart
Specifications
Chemical Name or Material
Potassium Phosphate Monobasic
Name Note
Anhydrous
CAS
7778-77-0
Recommended Storage
Room Temperature
Physical Form
Crystalline
View more