Check Order Status
Percoll™, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Percoll™, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219536990
500 mL
Catalog number
MPB0219536990
Price (SGD)
1,476.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
1,476.00
EA
Add to cart
Specifications
CAS
65455-52-9
Quantity
500 mL