Check Order Status
MES, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
MES, >98%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219530991
1 kg
Catalog number
MPB0219530991
Price (SGD)
1,823.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
1,823.00
EA
Add to cart
Specifications
Chemical Name or Material
MES
CAS
145224-94-8
Color
White
Recommended Storage
Store at room temperature.
EINECS Number
249-259-4
View more
Biological buffer