Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
MP Biomedicals
Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
Guanosine-5'-triphosphate Disodium Salt, ≥95% is a substrate for RNA polymerases.
Have Questions?
Catalog NumberQuantity
MPB021952132525 mg
Catalog number MPB0219521325
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialGuanosine-5'-triphosphate disodium salt
Name NoteFor HPLC analysis
CAS36051-31-7
Absorbanceλ max (water): 252 ± 3 nm
ColorWhite
View more
  • Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exchange factors (GEF).
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.