Check Order Status
Carbenicillin disodium salt, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Carbenicillin disodium salt, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219509205
5 g
Catalog number
MPB0219509205
Price (SGD)
679.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
679.00
EA
Add to cart
Specifications
Chemical Name or Material
Carbenicillin disodium salt
Type
Carbenicillin Disodium Salt
Physical Form
Powder
CAS
4800-94-6
Color
Yellow
View more