Check Order Status
Doxycycline hydrochloride, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Doxycycline hydrochloride, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219504401
1 g
Catalog number
MPB0219504401
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
Doxycycline hydrochloride
Type
Doxycycline Hydrochloride
Physical Form
Powder
CAS
10592-13-9
Color
Yellow
View more