Check Order Status
Amphotericin B, MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Amphotericin B, MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219504383
250 mg
Catalog number
MPB0219504383
Price (SGD)
250.00
EA
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (SGD)
250.00
EA
Add to cart
Specifications
Chemical Name or Material
Amphotericin B
Type
Amphotericin B
Name Note
from
Streptomyces sp.
Physical Form
Powder
CAS
1397-89-3
View more