Dispase
Product Image
MP Biomedicals
Dispase
From Bacillus polymyxa
Have Questions?
Catalog NumberQuantity
MPB02195022055 U
Catalog number MPB0219502205
Price (SGD)
150.00
EA
Add to cart
Quantity:
5 U
Price (SGD)
150.00
EA
Add to cart
Microbial metalloenzyme used in tissue dissociation (primary cell cultures) and subcultivation (secondary cell cultures). Extremely stable. Activity: 6U/mg. Unit Definition: One unit is the amount of enzyme which liberates 1.0μmole of Tyr per minute at 37°C, pH 7.5. MW: 35.9kDa. Lyophilized.
Ref.: Akimoto, Y., et al., J. Histochem. Cytochem., 40: 1129 (1992).
Specifications
ColorTan
pH6 to 8.5
For Use With (Application)Protein sequencing
SourceBacillus polymyxa
Quantity5 U
Product TypeDispase
Molecular Weight (g/mol)35.9
InhibitorsEDTA, EGTA, Hg2+, other heavy Metals
FormLyophilized Powder
Unit SizeEA
Contents & Storage
4°C