Check Order Status
Sodium citrate, 99.8%, For ACS analysis, MP Biomedicals™
Structure Search
MP Biomedicals
Sodium citrate, 99.8%, For ACS analysis, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219486891
1 kg
Catalog number
MPB0219486891
Price (SGD)
147.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
147.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium citrate
Name Note
For ACS analysis
CAS
1545801
Color
White
Molecular Formula
C6H9Na3O9
View more