Check Order Status
Triton™ X-100, Molecular Biology Reagent Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Triton™ X-100, Molecular Biology Reagent Grade, MP Biomedicals™
Have Questions?
Change view
Quantity:
250 mL
100 mL
Catalog number
MPB0219485483
Price (SGD)
158.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (SGD)
158.00
EA
Add to cart
Specifications
Chemical Name or Material
Triton™ X-100
Color
Colorless to Yellow
CAS
9002-93-1
Physical Form
Viscous Liquid
Recommended Storage
Store at room temperature (15° to 30°C)
View more