Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
MP Biomedicals
Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals™
Used in a wide variety of biochemical applications. Sodium Chloride, Molecular Biology Reagent Grade, MP Biomedicals is an essential nutrient found widely in nature. It is used as a buffer component, in precipitation and chemical sequencing of DNA, in chr
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Catalog number MPB0219484801
Price (SGD)
116.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
116.00
EA
Add to cart
Specifications
CAS7647-14-5
Recommended StorageStore at Room Temperature (15–30°C).
Physical FormCrystalline Powder
Quantity1 kg
Molecular FormulaClNa
View more
Sodium chloride is used in a wide variety of biochemical applications, including:
  • Intravenous fluids (normal saline is 0.9% w/v in water10)
  • Density gradients
  • A diluent to increase ionic strength in buffers or culture media
  • In salt-out procedures in the isolation of DNA
  • In high concentrations for preservation of foods, since bacteria cannot grow in high salt conditions
A salt-ice mixture in the ratio of approximately 33g sodium chloride to 100g ice (at −1°C) will drop in temperature to as low as −21°C, depending on the rate of stirring and the size of the ice chunks. Excess amounts of sodium chloride can destroy electrolyte balance and cause death in most animals, including humans.