Check Order Status
Polyethylene glycol, MP Biomedicals™
Structure Search
MP Biomedicals
Polyethylene glycol, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219483901
1 kg
Catalog number
MPB0219483901
Price (SGD)
179.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
179.00
EA
Add to cart
Specifications
Chemical Name or Material
Polyethylene glycol
Color
White
CAS
25322-68-3
Physical Form
Powder
Molecular Formula
(C2H4O)n
View more