Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
Polyethylene Glycol, MP Biomedicals™
MP Biomedicals
Polyethylene Glycol, MP Biomedicals™
Also known as PEG. Polyethylene Glycol, MP Biomedicals is a condensation polymer of ethylene oxide and water. It is used in precipitation of proteins, separating and purifying biomolecules, induction of cell hybridization, and as a fusing agent in enhanci
Have Questions?
Catalog NumberQuantity
MPB02194839.5500 g
Catalog number MPB02194839.5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
CAS25322-68-3
Physical FormWhite Prills
Recommended StorageStore at Room Temperature (15–30°C).
Molecular Formula(C2H4O)n
MDL NumberMFCD01779601
View more
PEGs are susceptible to oxidative degradation in the presence of air. Minimizing the exposure of PEG to elevated temperatures and/or exposure to oxygen, or addition of an antioxidant can limit the amount of degradation. PEGs do not hydrolyze or deteriorate upon storage. PEGs do not support the growth of molds.