Check Order Status
PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
PIPES, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219483880
100 g
Catalog number
MPB0219483880
Price (SGD)
207.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
207.00
EA
Add to cart
Specifications
Chemical Name or Material
PIPES
CAS
5625-37-6
Color
White
EINECS Number
227-057-6
Physical Form
Powder
View more