Check Order Status
Sodium dodecyl sulfate, 99.5%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sodium dodecyl sulfate, 99.5%, MP Biomedicals™
Have Questions?
Change view
Quantity:
1 kg
500 g
Catalog number
MPB0219483191
Price (SGD)
767.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
767.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium dodecyl sulfate
Color
White
CAS
151-21-3
Physical Form
Granular Powder
Quantity
1 kg
View more