Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
MP Biomedicals
Glycine, Molecular biology reagent, ≥99%, MP Biomedicals™
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
Catalog number MPB0219482594
Price (SGD)
564.00
EA
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (SGD)
564.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycine
CAS56-40-6
Recommended StorageStore at room temperature (15° to 30°C).
EINECS Number200-272-2
Assay Percent Range≥99%
View more
Protein Electrophoresis & Blotting