Check Order Status
D-(+)-Trehalose dihydrate, >99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Trehalose dihydrate, >99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219475680
100 g
Catalog number
MPB0219475680
Price (SGD)
549.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
549.00
EA
Add to cart
Specifications
Chemical Name or Material
D-(+)-Trehalose dihydrate
CAS
6138-23-4
Color
White
Recommended Storage
Store at Room Temperature (15 to 30°C).
EINECS Number
202-739-6
View more