Check Order Status
Sucrose, 99.97%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sucrose, 99.97%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB02194747.5
500 g
Catalog number
MPB02194747.5
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material
Sucrose
CAS
57-50-1
Color
White
Recommended Storage
Store at room temperature.
Density
1.587g/cm
3
at 25°C
View more