Check Order Status
L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
L-Proline, Cell Culture Reagent, ≥99%, MP Biomedicals™
L-Proline, Cell Culture Reagent, ≥99% is used in cell culture media and as a component of MEM non-essential amino acids solution.
Have Questions?
Catalog Number
Quantity
MPB0219472825
25 g
Catalog number
MPB0219472825
Price (SGD)
82.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
82.00
EA
Add to cart
Specifications
Chemical Name or Material
L-Proline
Melting Point
221°C
Name Note
Hydroxy-L-proline free
CAS
147-85-3
Recommended Storage
Room temperature
View more
L-Proline is used in cell culture media.
L-Proline is also a component of MEM non-essential amino acids solution.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.