Tween™ 80, MP Biomedicals™
Tween™ 80, MP Biomedicals™
Actual product may vary
Tween™ 80, MP Biomedicals™
Tween™ 80, MP Biomedicals™
Tween™ 80, MP Biomedicals™
MP Biomedicals
Tween™ 80, MP Biomedicals™
Tween™ 80 is widely used in a number of biochemical applications.
Have Questions?
Catalog NumberQuantity
MPB02194725.1100 mL
Catalog number MPB02194725.1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Tweens™ are a series of nonionic surfactants derived from sorbitan esters.
  • They are soluble or dispersible in water but differ widely in organic and oil solubilities.
  • Used as oil-in-water emulsifiers in pharmaceuticals, cosmetics, cleaning compounds, etc.
  • Polysorbate 20 is nonionic surfactants derived from sorbitan ester.
Specifications
DescriptionTween™ 80
CAS9005-65-6
SynonymPolyoxyethylenesorbitan monooleate; POE (20) sorbitan monooleate; Polyethylene glycol sorbitan monooleate; Polysorbate 80
FormLiquid
For Use With (Application)Selective protein extraction; Isolation of nuclei from mammalian cell lines
Quantity100 mL
Unit SizeEA
Contents & Storage
Store at Room Temperature