Check Order Status
Polysorbate 20, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Polysorbate 20, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219472404
4 L
Catalog number
MPB0219472404
Price (SGD)
800.00
EA
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (SGD)
800.00
EA
Add to cart
Specifications
Chemical Name or Material
Polysorbate 20
Color
Yellow
CAS
9005-64-5
Physical Form
Oily Liquid
Density
1.095 g/mL
View more