Check Order Status
D-(+)-Glucose, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Glucose, MP Biomedicals™
D-(+)-Glucose Anhydrous, Cell Culture Reagent, White Powder
Have Questions?
Catalog Number
Quantity
MPB02194672.1
100 g
Catalog number
MPB02194672.1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
CAS
50-99-7
Melting Point
150°C to 170°C (literature)
Storage Note 1
Store at Room Temperature (15-30°C).
Color
White
Density
1.544g/mL
View more
D-(+)-Glucose, anhydrous is a main source of energy for living organisms.
It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.
D-(+)-Glucose is a monosaccharide. It is a mixture of α- and β-anomers, primarily the α-anomer.