Check Order Status
Choline chloride, cell culture reagent, ≥99%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Choline chloride, cell culture reagent, ≥99%, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219463990
500 g
Catalog number
MPB0219463990
Price (SGD)
225.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
225.00
EA
Add to cart
Specifications
Chemical Name or Material
Choline chloride
CAS
67-48-1
Color
White
Density
1.1g/cm
3
at 20°C
Molecular Formula
C5H14ClNO
View more