Check Order Status
N-Acetyl-L-cysteine (Crystalline), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
N-Acetyl-L-cysteine (Crystalline), MP Biomedicals
Have Questions?
Change view
Quantity:
25 g
5 g
Catalog number
MPB0219460325
Price (SGD)
143.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (SGD)
143.00
EA
Add to cart
Specifications
Chemical Name or Material
N-Acetyl-L-cysteine
CAS
616-91-1
Color
White
EINECS Number
210-498-3
Assay Percent Notes
≥96%
View more