Nigrosin Stain
Nigrosin Stain
Actual product may vary
MP Biomedicals
Nigrosin Stain
Used as a viable cell staining agent especially in stem cell biology
Have Questions?
Catalog NumberQuantity
MPB0219459380100 g
Catalog number MPB0219459380
Price (SGD)
236.00
EA
Add to cart
Quantity:
100 g
Price (SGD)
236.00
EA
Add to cart
  • Formula Weight: 737.87
  • CAS # 8005-03-6
  • Synonym: Acid Black 2; C.I. 50420
  • Physical Description: Black crystalline powder
  • Solublility: Soluble in water(1mg/ml-opaque black with purple cast solution)
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
TypeNigrosin
Quantity100 g
FormatCrystal
For Use With (Application)Viable cell staining agent especially in stem cell biology
Storage Requirements15 to 30°C
Unit SizeEA