Check Order Status
Hormones: Hydrocortisone
Actual product may vary
MP Biomedicals
Hormones: Hydrocortisone
Have Questions?
Catalog Number
Quantity
MPB0219456901
1 g
Catalog number
MPB0219456901
Price (SGD)
83.00
EA
Add to cart
Quantity:
1 g
Price (SGD)
83.00
EA
Add to cart
For research use only. Not intended for therapeutic use.
Specifications
Product Type
Hydrocortisone
Sterility
Sterile
Quantity
1 g
Source
Soy beans
Unit Size
EA
Contents & Storage
15° to 30°C