Aprotinin
Aprotinin
Actual product may vary
MP Biomedicals
Aprotinin
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB0219455980100 mg
MPB021945592525 mg
MPB021945591010 mg
Catalog number MPB0219455980
Price (SGD)
2,186.00
EA
Add to cart
Quantity:
100 mg
Price (SGD)
2,186.00
EA
Add to cart
Cell dissociation reagent isolated from bovine lung. Lyophilized.
Specifications
FormPowder
Product TypeTrypsin inhibitor
SterilitySterile
pH5 to 7
Quantity100 mg
SourceBovine lung
Unit SizeEA
Contents & Storage
4°C