Tris Hydrochloride, MP Biomedicals™
Tris Hydrochloride, MP Biomedicals™
Tris Hydrochloride, MP Biomedicals™
Tris Hydrochloride, MP Biomedicals™
MP Biomedicals
Tris Hydrochloride, MP Biomedicals™
Tris-HCl is excellent biochemical and biological buffer for all applications where ultra high purity is required.
Have Questions?
Catalog NumberQuantity
MPB02194558945 kg
Catalog number MPB0219455894
Price (SGD)
2,621.00
EA
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (SGD)
2,621.00
EA
Add to cart
Specifications
CAS1185-53-1
Recommended StorageStore at Room Temperature (15–30°C)
Physical FormCrystalline Powder
Quantity5 kg
Molecular FormulaC4H12ClNO3
View more
The pH values of all buffers are temperature and concentration dependent.
  • For Tris buffers, pH increases about 0.03 unit per°C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
  • For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.