▸
Buffers and Standards
▸
Life Science Buffers
▸
TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
TRIS hydrochloride, >99%, Cell culture reagent grade, MP Biomedicals™
Excellent biochemical and biological buffer for all applications where ultra high purity is required. The pH values of all buffers are temperature and concentration dependent
Have Questions?
Change view
Quantity:
100 g
500 g
1 kg
Catalog number
MPB0219455880
Price (SGD)
116.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (SGD)
116.00
Each
Add to cart
Specifications
Chemical Name or Material
TRIS hydrochloride
CAS
1185-53-1
Color
White
EINECS Number
214-684-5
Physical Form
Crystalline Powder
View more