▸
Buffers and Standards
▸
Life Science Buffers
▸
HEPES, >98%, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
HEPES, >98%, MP Biomedicals™
Have Questions?
Change view
Quantity:
1 kg
10 g
25 g
50 g
100 g
250 g
500 g
Catalog number
MPB0219454991
Price (SGD)
1,207.00
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
1,207.00
Each
Add to cart
Specifications
Chemical Name or Material
HEPES
CAS
7365-45-9
Color
White
Recommended Storage
Store at Room Temperature (15 to 30°C). Store Desiccated.
EINECS Number
230-907-9
View more