Penicillin G, Sodium Salt, MP Biomedicals™
Penicillin G, Sodium Salt, MP Biomedicals™
MP Biomedicals
Penicillin G, Sodium Salt, MP Biomedicals™
Penicillin G, Sodium Salt Cell culture grade, Powder
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mU
25 mU
Catalog number MPB0219453780
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mU
Request bulk or custom format
Specifications
Physical FormPowder
Storage Note 1Store at +4°C, protect from light.
Recommended Storage4°C
Loss on Drying0.001
RTECS NumberXH9800000
View more
  • Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae.
  • It has been used to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility and in cell culture both alone and combined with streptomycin and other antibiotics.
  • Penicillin G also known as benzylpenicillin.
  • It exert a bacteriocidal as well as a bacteriostatic effect on susceptible bacteria by interfering with the final stages of the synthesis of peptidoglycan, a substance in the bacterial cell wall.