Antibiotics: Mitomycin
Antibiotics: Mitomycin
Actual product may vary
MP Biomedicals
Antibiotics: Mitomycin
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Have Questions?
Catalog NumberQuantity
MPB02194532022 mg
Catalog number MPB0219453202
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 mg
  • Format: Powder, Non-sterile
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Specifications
FormBlue powder
Product TypeAntibiotic
Quantity2 mg
Unit SizeEA
Contents & Storage
4°C