Kanamycin Sulfate
Kanamycin Sulfate
Actual product may vary
MP Biomedicals
Kanamycin Sulfate
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB021945312525 g
MPB02194531055 g
Catalog number MPB0219453125
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Antibiotic effective against most gram positive and gram negative bacteria and most types of yeast.
Specifications
FormCrystalline powder
Product TypeAntibiotic
Quantity25 g
Unit SizeEA
Contents & Storage
15° to 30°C