Gentamicin Sulfate
Gentamicin Sulfate
Actual product may vary
MP Biomedicals
Gentamicin Sulfate
Serve variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB02194530055 g
MPB02194530011 g
Catalog number MPB0219453005
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
General antimicrobial effective against gram positive bacteria, gram negative bacteria, and mycoplasma.
Specifications
FormPowder
Product TypeAntibiotic
SterilityNon-sterile
pH3.5 to 5.5
Quantity5 g
Unit SizeEA
Contents & Storage
4°C