Ampicillin, Sodium Salt
Ampicillin, Sodium Salt
Actual product may vary
MP Biomedicals
Ampicillin, Sodium Salt
Serves a variety of purposes in cell culture, including selection, elimination of mycoplasma and microbial contamination, and prophylaxis
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
MPB021945262525 g
MPB02194526055 g
Catalog number MPB0219452625
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
All antibiotics are intended for research use only. They are not for therapeutic use in humans or animals.
Applications:
Selection antibiotic effective against most gram negative and gram positive bacteria.
Specifications
FormPowder
Product TypeAntibiotic
SterilityNon-sterile
pH8 to 10
Quantity25 g
Unit SizeEA
Contents & Storage
4°C