Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Cell Culture Media, Supplements, and Reagents
▸
Cell Culture Reagents, Antibiotics and Supplements
▸
Antibiotics
▸
Hygromycin B
Actual product may vary
MP Biomedicals
Hygromycin B
Research use only
Have Questions?
Catalog Number
Quantity
MPB0219417083
250 mg
Catalog number
MPB0219417083
Price (SGD)
256.00
Each
Add to cart
Quantity:
250 mg
Price (SGD)
256.00
Each
Add to cart
Molecular Formula: C
20
H
37
N
3
O
13
Molecular Weight: 527
Purity: 95%
Specifications
Form
Powder
Product Type
Antibiotic
Quantity
250 mg
Unit Size
Each
Contents & Storage
4°C