Check Order Status
Hydrochloric Acid, 36.5-38%, (ACS Reagent Grade), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Hydrochloric Acid, 36.5-38%, (ACS Reagent Grade), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219405491
1 L
Catalog number
MPB0219405491
Price (SGD)
153.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
153.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrochloric Acid
CAS
7647-01-0
Recommended Storage
Room Temperature
Assay Percent Range
36.5-38%
Quantity
1 L
View more