Glycine, MP Biomedicals™
Glycine, MP Biomedicals™
Glycine, MP Biomedicals™
Glycine, MP Biomedicals™
MP Biomedicals
Glycine, MP Biomedicals™
Glycine, ACS Reagent Grade, White Crystalline Powder
Have Questions?
Catalog NumberQuantity
MPB02194049911 kg
Catalog number MPB0219404991
Price (SGD)
754.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
754.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycine
Melting Point240°C (Decomposition)(Literature)
Name NoteFor ACS analysis
CAS56-40-6
ColorWhite
View more
  • It is widely used as a buffer for a variety of immunological applications.
  • It is also used as an eluent in column chromatography.
  • It has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
  • Commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots , a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
  • Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon.
  • It is a major inhibitory neurotransmitter.