Check Order Status
Formaldehyde ACS MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Formaldehyde ACS MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0219404790
500 mL
Catalog number
MPB0219404790
Price (SGD)
128.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
128.00
EA
Add to cart
Specifications
Chemical Name or Material
Formaldehyde, ACS
Name Note
Contains 10-15% methanol.
CAS
50-00-0
Density
1.03 (air equals 1)
Assay Percent Range
36.5-38%
View more