Check Order Status
D-(+)-Glucose, Molecular biology reagent grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
D-(+)-Glucose, Molecular biology reagent grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219402491
1 kg
Catalog number
MPB0219402491
Price (SGD)
202.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (SGD)
202.00
EA
Add to cart
Specifications
Chemical Name or Material
D-(+)-Glucose
CAS
50-99-7
Color
White
Recommended Storage
Store at room temperature.
Density
1.544g/mL
View more