Check Order Status
Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Sucrose, 99% max., Molecular biology reagent grade, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219401890
500 g
Catalog number
MPB0219401890
Price (SGD)
85.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (SGD)
85.00
EA
Add to cart
Specifications
Chemical Name or Material
Sucrose
CAS
57-50-1
Color
White
Recommended Storage
Store at room temperature.
Density
1.587g/cm
3
at 25°C
View more