Check Order Status
Isopropyl alcohol, ≥99%, Reagent grade, For ACS analysis, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Isopropyl alcohol, ≥99%, Reagent grade, For ACS analysis, MP Biomedicals™
Have Questions?
Catalog Number
Quantity
MPB0219400791
1 L
Catalog number
MPB0219400791
Price (SGD)
137.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (SGD)
137.00
EA
Add to cart
Specifications
Chemical Name or Material
Isopropyl alcohol
Name Note
For ACS analysis
CAS
67-63-0
Color
Colorless
Density
0 .784 to 0.786 g/cm
3
at 20°C
View more